Skip to Content
Merck
All Photos(1)

Key Documents

CRM74120

Supelco

Triazophos

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€210.00

€210.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€210.00

About This Item

Empirical Formula (Hill Notation):
C12H16N3O3PS
CAS Number:
Molecular Weight:
313.31
Beilstein:
682554
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€210.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

−20°C

SMILES string

CCOP(=S)(OCC)Oc1ncn(n1)-c2ccccc2

InChI

1S/C12H16N3O3PS/c1-3-16-19(20,17-4-2)18-12-13-10-15(14-12)11-8-6-5-7-9-11/h5-10H,3-4H2,1-2H3

InChI key

AMFGTOFWMRQMEM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Triazophos― a non-systemic insecticide and acaricide, is an acetylcholinesterase (AChE) inhibitor. An organophosphate, it covalently and irreversibly reacts with the acetylcholine binding site and blocks the hydrolysis of acetylcholine, resulting in hyper excitation. Its route of action includes both ingestion and contact. Triazophos is used against soil insects and mites such as aphids, thrips, midges, beetles, Lepidoptera larvae, cutworms, and spider mites in ornamentals, cotton, rice, maize, soybeans, oil palms, olives, and coffee.
As per Regulation (EC) No. 1107/2009, repealing directive 91/414/EEC, triazophos is not approved for use in the European Union as a plant protection agent. Maximum residue limits (MRLs) applied for the presence of triazophos in food products across different countries lie in the range of 0.01 to 0.2 mg/kg.
Triazophos has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Triazophos CRM may find the following uses:

  • Analysis of a diagnostic method for the direct monitoring of triazophos by a non-competitive surface plasmon resonance immunosensor in environmental and agricultural product samples
  • Simultaneous quantitative detection of 171 pesticide residues using multi-walled carbon nanotubes (MWCNTs) as a sorbent in the reversed-dispersive solid-phase (r-DSPE) extraction of the cowpea samples followed by gas chromatographic-tandem mass spectrometric (GC-MS/MS) analysis
  • Development and validation of a multi-residue method for the analysis of 160 pesticides in herbal plants samples of chamomile, thyme, and marjoram using ethyl acetate and n-hexane mixture for the extraction before GC-MS/MS direct injection
  • Determination of a wide group of 250 pesticide residues in surface water samples by solid-phase extraction (SPE) and reversed-phase liquid chromatography (LC)-Orbitrap high-resolution mass spectrometry (HRMS)
  • Quantifying 126 pesticide residues in chicken muscle samples using QuEChERS for sample preparation and ultra-high-performance liquid chromatography- quadrupole time-of-flight-mass spectrometry (UHPLC-QToF-MS) for subsequent analysis

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Casado et al.
Analytica chimica acta, 1024, 1-17 (2018-05-20)
A quantitative targeted screening method for the determination of residues of a broad group of more than 250 pesticides in surface water samples was developed and validated. Substances were isolated from the sample matrix by solid-phase extraction (SPE), using hydrophilic-lipophilic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service