Skip to Content
Merck
All Photos(1)

Key Documents

CRM68817

Supelco

Bitertanol

certified reference material, TraceCERT®, mixture of stereo isomers, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

3,3-Dimethyl-1-(4-phenylphenoxy)-1-(1,2,4-triazol-1-yl)butan-2-ol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€129.00

€129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€129.00

About This Item

Empirical Formula (Hill Notation):
C20H23N3O2
CAS Number:
Molecular Weight:
337.42
Beilstein:
620948
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€129.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(C)C(O)C(Oc1ccc(cc1)-c2ccccc2)n3cncn3

InChI

1S/C20H23N3O2/c1-20(2,3)18(24)19(23-14-21-13-22-23)25-17-11-9-16(10-12-17)15-7-5-4-6-8-15/h4-14,18-19,24H,1-3H3

InChI key

VGPIBGGRCVEHQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Bitertanol is a chiral triazole fungicide used as a foliar spray or seed treatment for its curative and protective action against scab, Monilinia spp, rusts, powdery mildew, black spot, leaf spot, and other diseases for different fruits, vegetables, ornamentals, and cereals. It is a non-systemic fungicide that acts as an ergosterol biosynthesis inhibitor by targeting the activity of C14-demethylase and preventing the demethylation of sterol.
The approval for the use of bitertanol within European Union (EU) member states was withdrawn by means of Commission Implementing Regulation (EU) No 767/2013. But a default maximum residue limit of 0.01 mg/kg is applicable for the presence of bitertanol in plant protection products according to the Regulation (EC) No 396/200.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Bitertanol CRM may also be used as given below:

  • Separation and determination of bitertanol by ultra-performance liquid chromatography/tandem mass spectrometry (UPLC-MS/MS) to study its stereoselective degradation in tomato samples
  • Analysis of four stereoisomers of bitertanol in apple, pear, tomato, cucumber, and soil samples using high-performance liquid chromatography (HPLC)
  • Multiresidue determination of 133 pesticides in commercially available chenpi samples by a combination of modified QuEChERS method and gas chromatography-tandem mass spectrometry (GC-MS/MS)
  • Development of a method to determine eight triazole fungicides and their enantiomers in tomatoes, apples, peppers, eggplants, cucumber, and Chinese cabbage samples using aqueous two-phase extraction (ATPE) and two-dimensional liquid chromatography (2D-LC)
  • Using a modified QuEChERS method to extract 30 pesticides from raw bovine milk samples including a clean-up founded on dispersive solid-phase extraction (d-SPE) and the use of primary secondary amine (PSA) and zirconium dioxide (ZrO2)-based sorbents for the determination by high-performance liquid chromatography with diode-array detection (HPLC-DAD)
  • Simultaneous analysis of 20 triazole fungicides and plant growth regulators in cucumber samples by using molecularly imprinted polymers (MIPs) as a sorbent during the solid phase extraction (SPE), followed by high-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS)

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service