Skip to Content
Merck
All Photos(1)

Key Documents

CRM67607

Supelco

Metalaxyl Metabolite CGA 108906

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(±)-2-[(1-Carboxyethyl)(methoxyacetyl)amino]-3-methylbenzoic acid, N-(2-Carboxy-6-methylphenyl)-N-(methoxyacetyl)-DL-alanine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€153.00

€153.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€153.00

About This Item

Empirical Formula (Hill Notation):
C14H17NO6
CAS Number:
Molecular Weight:
295.29
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€153.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Metalaxyl Metabolite CGA 108906 is a benzoic acid derivative of the fungicide metalaxyl, formed upon its oxidation.
The Reg. (EU) 2017/1164 sets a maximum residue limit (MRL) of 0.7 mg/kg and 0.01 mg/kg in fresh/frozen fruits and tree nuts respectively for metalaxyl that includes its other mixtures of constituent isomers including metalaxyl-M (sum of isomers). Metalaxyl has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
The metalaxyl Metabolite CGA 108906 CRM may also find the following uses:

  • Analysis of insecticide and fungicide residues in soil samples by pressurized liquid extraction (PLE) accompanied by liquid chromatography-tandem mass spectrometry (LC-MS/MS) determination
  • Development of a multi-residue method for the determination of 150 pesticide metabolites in ground and surface water by direct injection-liquid chromatography-electrospray ionization-tandem mass spectrometry (DI-LC-ESI-MS/MS)

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thorsten Reemtsma et al.
Journal of chromatography. A, 1271(1), 95-104 (2012-12-12)
Based on the information available on 293 pesticides (herbicides, insecticides, fungicides, biocides, growth regulators) 210 pesticide metabolites were selected for inclusion into a multimethod for the analysis of ground and surface water. With the final method 150 pesticide metabolites can
L Pérez-Mayán et al.
Chemosphere, 261, 127696-127696 (2020-07-28)
The presence of fungicide and insecticide residues in wine has been largely investigated. However, few studies have addressed the persistence of these compounds in vineyard soils. In this research, we investigate the residues of a relevant number of fungicides and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service