Skip to Content
Merck
All Photos(1)

Key Documents

BP1189

Fluvastatin impurity F

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3-hydroxy-4,6-heptadienoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H24FNO3
CAS Number:
Molecular Weight:
393.45
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fluvastatin

form

crystalline solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

−20°C

SMILES string

Fc1ccc(cc1)c2c3c([n](c2\C=C\C=C\C(O)CC(=O)O)C(C)C)cccc3

InChI

1S/C24H24FNO3/c1-16(2)26-21-9-6-4-8-20(21)24(17-11-13-18(25)14-12-17)22(26)10-5-3-7-19(27)15-23(28)29/h3-14,16,19,27H,15H2,1-2H3,(H,28,29)/b7-3+,10-5+

InChI key

WARCEOCHTLFKTQ-GZTQOXHUSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluvastatin impurity F BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Fluvastatin Capsules
  • Fluvastatin Prolonged-release Tablets
  • Packaging

    Unit quantity: 25 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Storage Class Code

    11 - Combustible Solids

    WGK

    WGK 3

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service