Skip to Content
Merck
All Photos(1)

Key Documents

96355

Supelco

4,4′-Methylenebis(cyclohexylamine)

analytical standard

Synonym(s):

4,4′-Diaminodicyclohexylmethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(C6H10NH2)2
CAS Number:
Molecular Weight:
210.36
Beilstein:
2079179
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor pressure

<0.1 mmHg ( 38 °C)

Assay

≥94.0% mixed isomers basis (GC)

technique(s)

HPLC: suitable

density

0.95 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

NC1CCC(CC1)CC2CCC(N)CC2

InChI

1S/C13H26N2/c14-12-5-1-10(2-6-12)9-11-3-7-13(15)8-4-11/h10-13H,1-9,14-15H2

InChI key

DZIHTWJGPDVSGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

318.2 °F - closed cup

Flash Point(C)

159 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service