Skip to Content
Merck
All Photos(1)

Key Documents

93638

Supelco

Ethirimol

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€86.50

€86.50


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
50 MG
€86.50

About This Item

Empirical Formula (Hill Notation):
C11H19N3O
CAS Number:
Molecular Weight:
209.29
Beilstein:
882476
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€86.50


Estimated to ship on29 May 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

SMILES string

CCCCc1c(C)nc(NCC)nc1O

InChI

1S/C11H19N3O/c1-4-6-7-9-8(3)13-11(12-5-2)14-10(9)15/h4-7H2,1-3H3,(H2,12,13,14,15)

InChI key

BBXXLROWFHWFQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Ethirimol is the ethylamino isomer of dimethirimol that belongs to a group of pyrimidine fungicides. This systemic fungicide acts by inhibiting nucleic acid synthesis and is used as a seed treatment to control powdery mildew in spring barley and wheat. Ethirimol is not approved for use in European Union (EU) as per Regulation (EC) No 1107/2009. But a default maximum residue level (MRLs) from 0.01 mg/kg to 2 mg/kg is applied for products of plant and animal origin, according to Regulation (EU) 2021/1531.

Ethirimol has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
The Ethirimol CRM may also find following uses:
  • Simultaneous determination of the captan, captafol, carbendazim, chlorothalonil, ethirimol, folpet, metalaxyl, and vinclozolin from drinking, river, and estuarine water using solid-phase disk extraction followed by gas and liquid chromatography
  • Study the effects of commercial nonionic polyoxyethylene surfactants on the foliar penetration and movement of ethirimol and diclobutrazol from suspension formulations applied to wheat plants at different growth stages
  • Study the key ultrastructural responses of Erysiphe graminis in barley due to the effects of ethirimol and propiconazole fungicides
  • Development of QuEChERS-ultra performance liquid chromatography-tandem mass spectrometry analysis method for residual determination of thiophanate-methyl, carbendazim, and ethirimol in strawberry
  • Study the effect of pressurized fluid extraction (PFE) on the recovery of bupirimate and its degradation product, ethirimol from a range of soil types

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service