Skip to Content
Merck
All Photos(1)

Documents

91816

Supelco

HPTLC calibration mix

in methanol, analytical standard

Synonym(s):

High Performance TLC Calibration Standards

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

form

liquid

shelf life

limited shelf life, expiry date on the label

composition

9-fluorenol (1689-64-1), 1000 mg/L
guanosine (118-00-3), 500 mg/L
octrizole (3147-75-9), 1000 mg/L
paracetamol (103-90-2), 1000 mg/L
phthalimide (85-41-6), 2000 mg/L
sulisobenzone (4065-45-6), 1000 mg/L
thioxanthone (492-22-8), 10 mg/L
thymidine (50-89-5), 1000 mg/L

concentration

in methanol

refractive index

n20/D 1.328-1.334

density

0.791-0.799 g/mL at 20 °C

application(s)

food and beverages

storage temp.

−20°C

Other Notes

This universal HPTLC mix (UHM), provides laboratories with a single solution, applicable for HPTLC system suitability tests (SST) for a wide range of chromatographic systems, with different polarities and selectivities.

This product is suitable for the CAMAG HPTLC system suitability test.

Analyte

Description

    9-Hydroxyfluorene 1000 mg/L

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about our recently launched HPTLC calibration mix, developed in collaboration with CAMAG, that enables for an efficient and reliable universal system suitability testing (SST).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service