Skip to Content
Merck
All Photos(1)

Key Documents

91607

Supelco

Isazophos

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€129.00

€129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€129.00

About This Item

Empirical Formula (Hill Notation):
C9H17ClN3O3PS
CAS Number:
Molecular Weight:
313.74
Beilstein:
8145294
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€129.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

−20°C

SMILES string

CCOP(=S)(OCC)Oc1nc(Cl)n(n1)C(C)C

InChI

1S/C9H17ClN3O3PS/c1-5-14-17(18,15-6-2)16-9-11-8(10)13(12-9)7(3)4/h7H,5-6H2,1-4H3

InChI key

XRHGWAGWAHHFLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Isazophos is a broad-spectrum insecticide and nematicide belonging to the chemical class of organophosphates. Isazophos is an acetylcholinesterase inhibitor, that is used on a wide variety of crops such as bananas, citrus fruits, cotton, beet crops, maize, rice, and sugarcane to control turf and soil insect pests like grubs, com rootworms, root maggots, and wireworms.

According to Regulation (EC) 1107/2009, isazophos is not approved for use in European Union (EU). A default maximum residue level (MRLs) of 0.01 mg/kg has been set according to Art 18(1)(b) Reg 396 / 2005.

Application

It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Isazophos CRM may also find its use as described below:
  • To develop a GC/MS procedure utilizing selected ion monitoring (SIM) for quantification of isazophos in bovine liver, kidney, muscle, and fat at various times after exposure
  • To develop multiresidue method for quantification of 19 metabolites of organophosphorus pesticides, synthetic pyrethroids, selected herbicides, and DEET (N,N-diethyl-m-toluamide) in human urine using LC-MS
  • To determine the efficacy of isazophos against Japanese beetle (Popillia japonica Newman) grubs using two subsurface placement technology, high-pressure liquid injection (HPLI) and an adapted overseeder (OS), applied to golf course roughs

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2 Inhalation

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service