Skip to Content
Merck
All Photos(1)

Key Documents

91386

Supelco

Iprobenfos

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€190.00

€190.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€190.00

About This Item

Empirical Formula (Hill Notation):
C13H21O3PS
CAS Number:
Molecular Weight:
288.34
Beilstein:
1974687
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€190.00


Estimated to ship on31 May 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

CC(C)OP(=O)(OC(C)C)SCc1ccccc1

InChI

1S/C13H21O3PS/c1-11(2)15-17(14,16-12(3)4)18-10-13-8-6-5-7-9-13/h5-9,11-12H,10H2,1-4H3

InChI key

FCOAHACKGGIURQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Iprobenfos belongs to a group of phosphorothiolate fungicides with both curative and protective properties. It is effective against Pyricularia oryzae, Helminthosporium sigmoideum, Magnaporthe salvinii, and Rhizoctonia solani and is widely applied to control rice leaf and ear blast, stem rot, and rice sheath blight in rice. Iprobenfos is a systemic fungicide that hinders phosphatidylcholine biosynthesis through inhibition of methyltransferase, thereby, interfering with membrane structure.

According to Regulation (EC) 1107/2009, repealing Council Directive 91/414/EEC, iprobenfos is not approved for use in European Union (EU). A default MRL of 0.01 mg/kg is set according to Art 18(1)(b) Reg 396 / 2005.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
The iprobenfos CRM may also find following uses:
  • Sensitive detection of iprobenfos and edifenphos using gold nanoparticle, AuNP-based colorimetric multi-aptasensor from rice samples
  • To investigate light-induced catalytic degradation of iprobenfos fungicide using poly(3-octylthiophene-2,5-diyl) film by varying oxygen pressure of bubbling gases
  • To determine the variation in sensitivity of Magnaporthe oryzae isolates from different geographic areas towards edifenphos and iprobenfos fungicides
  • To study the response of zooplankton community against the application of simetryn and iprobenfos using natural zooplankton community in microcosm tanks
  • To evaluate and compare the virulence of Magnaporthe oryzae isolates sensitive and resistant to edifenphos and iprobenfos on rice plants treated with or without fungicides

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service