Skip to Content
Merck
All Photos(1)

Key Documents

91359

Supelco

Propyl 4-hydroxybenzoate sodium salt

analytical standard

Synonym(s):

Sodium propyl 4-hydroxybenzoate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€79.90

€79.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€79.90

About This Item

Empirical Formula (Hill Notation):
C10H11NaO3
CAS Number:
Molecular Weight:
202.18
Beilstein:
5183350
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€79.90


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

form

crystals

shelf life

limited shelf life, expiry date on the label

SMILES string

[Na+].CCCOC(=O)c1ccc([O-])cc1

InChI

1S/C10H12O3.Na/c1-2-7-13-10(12)8-3-5-9(11)6-4-8;/h3-6,11H,2,7H2,1H3;/q;+1/p-1

InChI key

IXMINYBUNCWGER-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service