Skip to Content
Merck
All Photos(1)

Key Documents

90556

Supelco

Imazamethabenz-methyl

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€98.10

€98.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€98.10

About This Item

Empirical Formula (Hill Notation):
C16H20N2O3
CAS Number:
Molecular Weight:
288.34
Beilstein:
8328238
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€98.10


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

SMILES string

COC(=O)c1ccc(C)cc1C2=NC(C)(C(C)C)C(=O)N2

InChI

1S/C16H20N2O3/c1-9(2)16(4)15(20)17-13(18-16)12-8-10(3)6-7-11(12)14(19)21-5/h6-9H,1-5H3,(H,17,18,20)

InChI key

FFCCBBNQPIMUJI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Imazamethabenz-methyl– an acetolactate synthase inhibitor, is a mixture of meta- and para-isomers (2:3) that belongs to a group of imidazolinone herbicides. This systemic herbicide is applied postemergence in winter cereals and other crops to control grasses and some dicotyledonous weeds. Imazamethabenz-methyl is effective against many of the economically important weeds such as Avena spp., Apera spica-venti, Poa trivialis, Raphanus raphanistrum, and Brassica spp. that infest wheat, barley, and rye. The herbicide is readily absorbed through the foliage and roots of sensitive weeds and is slowly translocated to the meristematic regions. It functions by inhibiting acetohydroxyacid synthase enzyme, which is responsible for the synthesis of branched-chain amino acids in plants.

According to Regulation (EC) 1107/2009, repealing Council Directive 91/414/EEC, imazamethabenz-methyl is not approved for use in European Union (EU). A default MRL of 0.01 mg/kg is set according to Art 18(1)(b) Reg 396 / 2005.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
The imazamethabenz-methyl CRM may also find following uses:
  • Study the photolysis of imazamox, imazapic, imazaquin, and imazamethabenz-methyl as functions of wavelength, pH, and natural organic matter in an aqueous solution
  • Comparative study of the extraction of imazamethabenz-methyl from agricultural soils, followed by its chromatographic determination using HPLC
  • Investigate the absorption and translocation of fenoxaprop-ethyl and imazamethabenz-methyl in wild oat plants grown under different conditions of temperature and light intensity using 14C tracer techniques
  • Simultaneous determination of p- and m-imazamethabenz-methyl enantiomers using a protein chiral selector based on a direct chiral HPLC method
  • To develop a multiresidue method for the determination of post-emergence herbicides in water samples by SPE coupled with HPLC/ESI/MS in positive ionization mode

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service