Skip to Content
Merck
All Photos(1)

Key Documents

90146

Supelco

2-tert-Butyl-6-(5-chloro-2H-benzotriazol-2-yl)-4-methylphenol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18ClN3O
CAS Number:
Molecular Weight:
315.80
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Assay

≥97.5% (HPLC)

form

powder

shelf life

limited shelf life, expiry date on the label

mp

144-147 °C (lit.)

storage temp.

room temp

SMILES string

Cc1cc(c(O)c(c1)C(C)(C)C)-n2nc3ccc(Cl)cc3n2

InChI

1S/C17H18ClN3O/c1-10-7-12(17(2,3)4)16(22)15(8-10)21-19-13-6-5-11(18)9-14(13)20-21/h5-9,22H,1-4H3

InChI key

OCWYEMOEOGEQAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service