Pricing and availability is not currently available.
Product Name
TSKgel® SuperHZM-H HPLC Column, phase polystyrene-divinylbenzene, L × I.D. 15 cm × 4.6 mm, 10 μm particle size
material
stainless steel
Agency
suitable for USP L21
product line
TSKgel®
manufacturer/tradename
Tosoh 19664
parameter
0.4 mL/min flow rate 10 bar max. pressure 80 °C temperature
technique(s)
HPLC: suitable
L × I.D.
15 cm × 4.6 mm
matrix
polystyrene-divinylbenzene
matrix active group
polystyrene-divinylbenzene phase
particle size
10 μm
separation technique
size exclusion (SEC)
General description
TSKgel® SuperHZM-H mixed-bed columns are packed with rigid, porous, polymer beads each having a varying mean pore diameter. They are used for the non-aqueous separation of organic-soluble polymers with high molecular weights of less than 400,000,000 Da. The THF shipping solvent may be replaced by a limited number of organic solvents.
Physical form
THF can be replaced with benzene, chloroform, toluene, xylene, dichloromethane, dichloroethane.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
TSKgel is a registered trademark of Tosoh Corporation
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.