807469
TOYOPEARL® HW-75F Bulk Media
phase hydroxylated methacrylate, bottle of 500 mL, 45 μm particle size
Synonym(s):
TOYOPEARL® HW-75 Size Exclusion Media
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
HW-75F
product line
TOYOPEARL®
form
slurry
packaging
bottle of 500 mL
parameter
3 bar max. pressure
technique(s)
HPLC: suitable
matrix
hydroxylated methacrylic polymer
matrix active group
hydroxylated methacrylate phase
particle size
45 μm
pore size
100,000-10,000,000 Da MW range
operating pH
2-13
separation technique
size exclusion (SEC)
General description
Toyopearl HW-75 resin is a hydroxylated methacrylic polymer bead for size exclusion chromatography of proteins between 500,000-50,000,000 Da.
Application
TOYOPEARL® media fractionate mixtures of proteins and other large molecular weight compounds over a wide size range. Applications include separating RNA from protein, resolving oligosaccharides by degree of polymerization, and isolating agglutinin while maintaining a high hemagglutination titer.
Specifications
Clean in place with 0.5 M NaOH or 0.1 M HCl.
Physical form
Shipped in 20% (v/v) ethanol.
Legal Information
Toyopearl is a registered trademark of Tosoh Corporation
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
WGK
WGK 1
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protein expression and purification, 82(1), 150-154 (2012-01-10)
We have cloned ansB (YPTB1411) gene from Yersinia pseudotuberculosis Q66CJ2 and constructed stable inducible expression system that overproduce L-asparaginase from Y. pseudotuberculosis (YpA) in Escherichiacoli BL21 (DE3) cells. For purification of YpA we used Q-Sepharose and DEAE-Toyopearl column chromatography. We
Fitoterapia, 83(1), 153-160 (2011-11-01)
Persimmon proanthocyanidin was fractionated on Toyopearl TSK-HW-50-F to yield a fraction with strong inhibition on the catalytic activity and edema-inducing activity and lethality of Chinese cobra PLA(2). Thiolysis suggested that the terminal units included C, EGCG and myricetin, and epicatechin
Carbohydrate research, 347(1), 161-163 (2011-11-22)
A method is developed for the preparation of D-rhamnose from an O-polysaccharide (OPS) isolated by mild acid hydrolysis of Azospirillum brasilense SR75 cell mass. After the OPS hydrolysis, D-rhamnose was recovered by gel-permeation chromatography on Toyopearl TSK HW-40 and was
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service