80201
Extractables and Leachables PE/PP mix for GC
certified reference material, TraceCERT®, 21 compounds, in tert-butyl methyl ether, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
concentration
in tert-butyl methyl ether
application(s)
pharmaceutical
format
multi-component solution
storage temp.
−20°C
General description
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
Preparation Note
Legal Information
Kit Components Only
- 1,3-Di-tert-butylbenzene
- 1-Octene
- Butylated hydroxyanisole
- 2,6-Di-tert-butylphenol
- 2,6-Di-tert-butyl-4-methylphenol
- 2-Ethylhexanoic acid
- 2-Methyldodecane
- 3-Methyl-4-methylene-2-hexanone
- Isovaleric acid
- 7,9-Di-tert-butyl-1-oxaspiro[4.5]deca-6,9-diene-2,8-dione
- Benzophenone
- 1-Decene
- Dibutyl phthalate
- Didodecyl 3,3′-thiodipropionate
- Docosane
- Eicosane
- Hexadecane
- Isophorone
- Octadecane
- ε-Caprolactam
- Ethylene glycol butyl ether
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
-18.4 °F - closed cup
Flash Point(C)
-28 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service