Skip to Content
Merck
All Photos(1)

Key Documents

80023455

TSKgel® Amide-80 (2 µm) HPLC Columns

L x I.D. 10 cm × 2.0 mm, carbamoyl phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709

Product Name

TSKgel® Amide 80 HPLC Column, phase carbamoyl, L × I.D. 10 cm × 2.0 mm, 2 μm particle size

material

stainless steel

Agency

suitable for USP L68

product line

TSKgel®

manufacturer/tradename

Tosoh 23455

parameter

400 bar max. pressure
50 °C

technique(s)

HPLC: suitable

L × I.D.

10 cm × 2.0 mm

surface area

450 m2/g

matrix

fully porous particle

matrix active group

carbamoyl phase

particle size

2 μm

operating pH range

2.0-7.5

separation technique

hydrophilic interaction (HILIC)

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This article evaluates different phase chemistries for stationary phases in Hydrophilic Interaction Liquid Chromatography (HILIC) and uses multivariate analysis for classification based on chemical modification.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service