Skip to Content
Merck
All Photos(1)

Documents

68992

Supelco

2-Nitrophenyl octyl ether

analytical standard

Synonym(s):

1-Nitro-2-octyloxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4O(CH2)7CH3
CAS Number:
Molecular Weight:
251.32
Beilstein:
2529905
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.508 (lit.)

bp

197-198 °C/11 mmHg (lit.)

density

1.04 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CCCCCCCCOc1ccccc1[N+]([O-])=O

InChI

1S/C14H21NO3/c1-2-3-4-5-6-9-12-18-14-11-8-7-10-13(14)15(16)17/h7-8,10-11H,2-6,9,12H2,1H3

InChI key

CXVOIIMJZFREMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

May be used as a plasticizer for cellulose triacetate membranes providing impermeability to metal cations, but allowing anion exchange.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service