Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
57690-U
RheFlex® PEEK Fittings
Long fitting set, 1/16 in. tubing
Synonym(s):
Polyether Ether Ketone Tube Fittings
About This Item
packaging
pkg of 5 ea
compatibility
1/16 in. tubing
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the Department of Transportation shipping information for this product?
1 answer-
Helpful?
-
-
What is the length of product 57690-U, Rheflex PEEK Fitting?
1 answer-
The length of Rheflex PEEK Fitting 57690-U is 33 mm, or 1.32 in.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service