57234-U
SPME LC Tips, C18
tray of 96 ea
Synonym(s):
SPME LC TIps
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
packaging
tray of 96 ea
fiber L
1 cm
matrix active group
C18 coating
application(s)
food and beverages
compatibility
for use with Pipettor or robotic liquid handling system
Looking for similar products? Visit Product Comparison Guide
General description
Low cost, micro-sample prep devices consisting of a coated microfiber held in place in a micropipette tip making it compatible with both multi-channel pipettors and robotic liquid handling systems. The tips are sold in a 96 well tray configuration.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service