Skip to Content
Merck
All Photos(1)

Documents

55474-U

Supelco

Supel QuE QuEChERS tube

PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible

Synonym(s):

QuEChERS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

product name

Supel QuE, PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible

material

polypropylene tube

Agency

AOAC® 2007.01

product line

Supel

composition

Discovery® DSC-18, 400 mg
Magnesium Sulfate, 1200 mg
Supelclean ENVI-Carb, 400 mg
Supelclean PSA, 400 mg

packaging

pk of 50

technique(s)

QuEChERS: suitable

centrifuge tube volume

15 mL , Shaker Compatible

matrix active group

C18 phase
PSA phase
carbon phase

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

General description

Dispersive SPE (dSPE), often referred to as the "QuEChERS" method (Quick, Easy, Cheap, Effective, Rugged, and Safe), is modern sample prep technique that is becoming increasingly popular in the area of multi-residue pesticide analysis in food and agricultural products.

Using the QuEChERS method, food/agricultural samples are first extracted with an aqueous miscible solvent (e.g., acetonitrile) in the presence of high amounts of salts (e.g., sodium chloride and magnesium sulfate) and/or buffering agents (e.g. citrate) to induce liquid phase separation and stabilize acid and base labile pesticides, respectively. Upon shaking and centrifugation, an aliquot of the organic phase is subjected to further cleanup using SPE. Unlike traditional methods using SPE tubes, in dispersive SPE, cleanup is facilitated by mixing bulk amounts of SPE (e.g., Supelclean PSA, ENVI-Carb, and/or Discovery DSC-18) with the extract. After sample cleanup, the mixture is centrifuged and the resulting supernatant can either be analyzed directly or can be subjected to minor further treatment before analysis.

Supelco carries a line of vials and centrifuge tubes containing pre-determined amounts of salts and SPE sorbents to support the most common method configurations used today.

Compatibility

Compatible with most QuEChERS shakers.

Legal Information

AOAC is a registered trademark of AOAC International
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany
Supel is a trademark of Sigma-Aldrich Co. LLC
Supelclean is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U Führer et al.
Analytical and bioanalytical chemistry, 354(3), 333-343 (1996-01-01)
Halogenated anisoles (methyl-phenyl ethers) appear to be ubiquitous organic trace compounds in the environment. An analytical method is presented for analyzing the altogether 134 congeners of chloro-, bromo- and mixed bromochloro-anisoles on an isomer-specific basis in air. High volume sampling
Chun-Lin Fan et al.
Journal of AOAC International, 98(1), 130-148 (2015-04-11)
A 3 month study was conducted on the ruggedness of a multiresidue method for accuracy and stability. The results indicate that in terms of Youden pair ratios of 201 pesticide aged tea samples falling approximately within 1.00-1.20 of the ratio
Tsuyoshi Imazawa et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 46(6), 277-281 (2006-01-31)
An analytical method was developed for the determination of phenmedipham (PM) in agricultural products using reversed-phase high-performance liquid chromatography with UV detection. A sample was extracted with acetonitrile, and the acetonitrile layer was separated by salting-out. The acetonitrile phase was
Sachiyo Ibuki et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 48(5), 139-143 (2007-11-22)
A method for simultaneous determination of Dichlorvos (DDVP), Trichlorfon (DEP) and Naled (BRP) in fruits and vegetables by liquid chromatography with tandem mass spectrometry (LC/MS/MS) was developed. Pesticides were extracted with ethyl acetate together with phosphoric acid and anhydrous sodium
Anna Martínez-Villalba et al.
Electrophoresis, 34(6), 870-876 (2013-01-22)
Veterinary medicines are widely administered to farm animals since they keep animals healthy at overcrowded conditions. Nevertheless the continuous administration of medicines to farm animals can frequently lead to the presence of residues of veterinary drugs in consumption products. Amprolium

Related Content

QuEChERS method simplifies sample cleanup for pesticides analysis in food and agricultural samples.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service