Skip to Content
Merck
All Photos(1)

Key Documents

55233-U

Supelco

Supel QuE

PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS

Synonym(s):

QuEChERS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

Agency

suitable for BS EN 15662:2008

composition

magnesium sulfate, 900 mg
Supelclean
ENVI-Carb, 45 mg (Cat. No. 57210-U)

Supelclean
PSA, 150 mg (Cat. No. 52738-U)

packaging

pk of 50

technique(s)

QuEChERS: suitable

centrifuge tube volume

12 mL , Greiner Cat. No. 163270

application(s)

food and beverages

separation technique

normal phase

Looking for similar products? Visit Product Comparison Guide

General description

Dispersive SPE (dSPE), often referred to as the "QuEChERS" method (Quick, Easy, Cheap, Effective, Rugged, and Safe), is modern sample prep technique that is becoming increasingly popular in the area of multi-residue pesticide analysis in food and agricultural products.

Using the QuEChERS method, food/agricultural samples are first extracted with an aqueous miscible solvent (e.g., acetonitrile) in the presence of high amounts of salts (e.g., sodium chloride and magnesium sulfate) and/or buffering agents (e.g. citrate) to induce liquid phase separation and stabilize acid and base labile pesticides, respectively. Upon shaking and centrifugation, an aliquot of the organic phase is subjected to further cleanup using SPE. Unlike traditional methods using SPE tubes, in dispersive SPE, cleanup is facilitated by mixing bulk amounts of SPE (e.g., Supelclean PSA, ENVI-Carb, and/or Discovery DSC-18) with the extract. After sample cleanup, the mixture is centrifuged and the resulting supernatant can either be analyzed directly or can be subjected to minor further treatment before analysis.

Supelco carries a line of vials and centrifuge tubes containing pre-determined amounts of salts and SPE sorbents to support the most common method configurations used today.

Application

Supel QuE may be used as a quick, easy, cheap, effective, rugged, safe extraction tube for the dispersive solid phase extraction of pesticide residues in vegetables using acetylcholinesterase (AChE) inhibition assay.[1]

Suitability

Suitable for food/agricultural samples with higher levels of chlorophyll and carotinoides (e.g., red sweet pepper, spinach, lamb′s lettuce, ruccolla, etc.). Ideal for clean up of 6 mL extract after initial food extraction and phase partitioning.

Legal Information

Supel is a trademark of Sigma-Aldrich Co. LLC
Supelclean is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Analysis of non-cleaned QuEChERS extracts for the determination of pesticide residues in fruit, vegetables and cereals by gas chromatography-tandem mass spectrometry
Norli RH, et al.
Food Additives & Contaminants. Part A, Chemistry, Analysis, Control, Exposure & Risk Assessment, 33(2), 300-312 (2016)
Tsuyoshi Imazawa et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 46(6), 277-281 (2006-01-31)
An analytical method was developed for the determination of phenmedipham (PM) in agricultural products using reversed-phase high-performance liquid chromatography with UV detection. A sample was extracted with acetonitrile, and the acetonitrile layer was separated by salting-out. The acetonitrile phase was
Anna Martínez-Villalba et al.
Electrophoresis, 34(6), 870-876 (2013-01-22)
Veterinary medicines are widely administered to farm animals since they keep animals healthy at overcrowded conditions. Nevertheless the continuous administration of medicines to farm animals can frequently lead to the presence of residues of veterinary drugs in consumption products. Amprolium
Sachiyo Ibuki et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 48(5), 139-143 (2007-11-22)
A method for simultaneous determination of Dichlorvos (DDVP), Trichlorfon (DEP) and Naled (BRP) in fruits and vegetables by liquid chromatography with tandem mass spectrometry (LC/MS/MS) was developed. Pesticides were extracted with ethyl acetate together with phosphoric acid and anhydrous sodium
Dajie Chen et al.
Se pu = Chinese journal of chromatography, 30(9), 896-902 (2013-01-05)
A high performance liquid chromatography-tandem mass spectrometry (HPLC-MS/ MS) method was established for the determination of fungicide anilinopyrimidine residues including pyrimethanil, mepanipyrim, and cyprodinil in foodstuffs with series solid phase extraction (SPE). The food sample was first extracted with ethyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service