Skip to Content
Merck
All Photos(1)

Documents

54842

Sigma-Aldrich

MAbTrap® Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

storage temp.

2-8°C

General description

For fast, effective purification of monoclonal and polyclonal IgG from ascites fluid, serum, or cell culture media, via syringe or pump. Sufficient materials for 20 purifications.

Application

MabTrap kits have been used in studies assessing physiological mechanisms of heart failure as well as to quantify des-Arg9-bradykinin.

Footnote

For fast, effective purification of monoclonal and polyclonal IgG from ascites fluid, serum, or cell culture media, via syringe or pump. Sufficient materials for 20 purifications. Contents: 1mL HiTrap protein G column, 50mL 10X binding buffer concentrate (0.05% sodium azide preservative), 15mL 10X elution buffer concentrate (0.05% sodium azide), 25mL neutralizing buffer (0.05% sodium azide), luer adapter, domed nut, syringe, instructions.

Legal Information

MAbTrap is a registered trademark of Cytiva

Kit Components Only

Product No.
Description

  • 10× binding buffer 50 mL

  • 10× elution buffer 15 mL

  • HiTrap Protein G column 1 mL

  • Neutralizing buffer 25 mL

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Distribution and property of anti-β3-adrenoceptor autoantibody in patients with heart failure
Li, M.X., et al.
Zhonghua Yi Xue Za Zhi, 33, 1114-1118 (2005)
P Raymond et al.
Journal of immunological methods, 180(2), 247-257 (1995-03-27)
There is a renewed interest in the kininase I pathway of kinin metabolism, because des-Arg9-bradykinin (des-Arg9-BK) and des-Arg10-Lys-BK are selective and potent agonists of the B1 receptors, that are apparently upregulated by tissue injury. We have developed a polyclonal rabbit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service