Skip to Content
Merck
All Photos(1)

Key Documents

52578-U

Supelco

Supelclean PSA SPE Tube

bed wt. 200 mg, volume 3 mL, pk of 54

Synonym(s):

PSA SPE cartridge tube, 3 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21
Pricing and availability is not currently available.

material

PE frit (20 μm porosity)
polypropylene hardware

product line

Supelclean

composition

bed wt., 200 mg

packaging

pk of 54

extent of labeling

8.4% C loading
3.3% N loading

technique(s)

solid phase extraction (SPE): suitable

surface area

500 m2/g

volume

3 mL

surface coverage

surface coverage 2.35 μmol/m2

matrix

silica gel base material (irregularly shaped)

matrix active group

ethylenediamine-N-propy bonding, polymerically bonded

particle size

50 μm

pore size

70 Å pore size

pKa 

(1) 10.1, (2) 10.9

capacity

0.98-1.05 meq/g capacity

application(s)

food and beverages

separation technique

ion exchange
normal phase

Looking for similar products? Visit Product Comparison Guide

General description

Retention Mechanism: Normal-phase and anion-exchange
Sample Matrix Compatibility: Organic or aqueous solutions

  • Polymerically bonded, ethylenediamine-N-propyl phase that contains both primary and secondary amines
  • A weak anion exchanger with a pKa of 10.1 and 10.9
  • Similar to aminopropyl SPE phases (NH2) in terms of selectivity, but has a much higher capacity due to presence of secondary amine (0.98-1.05 meq/g)
  • Strong affinity and high capacity for removing fatty acids, organic acids, and some polar pigments and sugars when conducting multi-residue pesticide analysis in foods
  • Has been shown to significantly reduce matrix-enhancement effects encountered during the GC analysis of food products
  • Tested for superior cleanliness using GC-FID and GC-MS
  • Bidentate nature of ligand allows for chelation

Legal Information

Supelclean is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

respiratory tract irritation

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service