Skip to Content
Merck
All Photos(1)

Key Documents

51172-U

OPTI-SOLV® Mini, Micro, and Nano Filter

micro filter, 1.0μm, pkg of 5 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

packaging

pkg of 5 ea

Looking for similar products? Visit Product Comparison Guide

General description

The OPTI-SOLV® Mini Filter provides low-impact filtering in a package no longer that a finger tight fitting. Use them to prolong the life of your analytical colum, or before your mass spectrometer as a last line of defense against debris.

The OPTI-SOLV® Micro Filter is based on the same design as the Mini Filter and cuts the internal volume to less that 200 nL. It retains the Mini Filter′s ease of use and functionality. The OPTI-SOLV® Micro Filter features a zero-dead volume connection utilizing Optimize Technologies® patented floating stem technology.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

OPTI-SOLV is a registered trademark of Optimize Technologies, Inc.
Optimize Technologies is a registered trademark of Optimize Technologies, Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service