Skip to Content
Merck
All Photos(1)

Key Documents

505293

Supelco

LpDNPH S10x Cartridge

bed wt. 350 mg, volume 1.5 mL, pkg of 10 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000
Pricing and availability is not currently available.

Agency

ASTM® D5197
EPA 100,IP-6A,TO-11A,TO-11A
NIOSH 2016

composition

bed wt., 350 mg
2 4-DNPH (High-purity silica gel)

packaging

pkg of 10 ea

technique(s)

active air sampling: suitable

volume

1.5 mL

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with XonTech 920/924, ATEC Atmospheric Technology Instruments

storage temp.

2-8°C

General description

LpDNPH S10x Cartridge is an air sampling device designed for sampling in ambient, indoor, and industrial atmospheres carbonyls. Carbonyls are trapped on a high purity silica adsorbent coated with 2,4-dinitrophenylhydrazine, where they are converted to the hydrazone derivatives. The derivatives are eluted from the cartridge in acetonitrile and are analyzed by HPLC.
The S10x cartridge is shorter than the S10 cartridge and designed to fit into automated instruments such as XonTech, Inc. and ATEC Atmospheric Technology. Using the S10x cartridge, the air sampling flow is in the direction opposite compared to the S10 cartridge in order to accommodate the instruments flow pattern.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service