Skip to Content
Merck
All Photos(1)

Key Documents

5028-U

Supelco

ASSET® EZ4 Dry Samplers

pkg of 50 ea

Synonym(s):

ASSET® EZ4-NCO Dry Sampler

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000
NACRES:
NB.24
Pricing and availability is not currently available.

product line

ASSET®

Quality Level

packaging

pkg of 50 ea

technique(s)

active air sampling: suitable

matrix

DBA-impregnated glass fiber filters

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for sampling isocyanate monomers & oligomers (*not recommended for isocyanic acid (ICA))
for use with LC-MS or LC-MS/MS

Looking for similar products? Visit Product Comparison Guide

General description

We offer ASSET® EZ4 Dry Samplers that are available in convenient packages of 50 units. They are compatible with sampling isocyanate monomers and oligomers and offer exceptional sensitivity for the collection and measurement of vapor phase and particulate isocyanates.

ASSET® EZ4 Dry Samplers are user-friendly, making them easy to use. They consistently deliver reliable results, which are useful in conjunction with LC-MS or LC-MS/MS analysis.

Application

ASSET® EZ4 Dry Samplers is used for the collection and monitoring of air samples in various industries, including environmental, occupational health, and safety assessments. [1]

Legal Information

Asset is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B Inhalation - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HBM4EU diisocyanates study?research protocol for a collaborative European human biological monitoring study on occupational exposure.
Kate J, et al.
International Journal of Environmental Research and Public Health, 19, 8811-8811 (2022)
Daniel Gylestam et al.
The Annals of occupational hygiene, 58(3), 340-354 (2013-12-29)
An advanced design of a denuder impactor (DI) sampler has been developed for characterization of possible airborne isocyanate exposure in different particle size fractions. The sampler is equipped with 12 different parallel denuder tubes, 4 impaction stages with the cut-off

Related Content

Easy-to-use air sampler offering ultimate sensitivity for collection and measurement of vapor phase and particulate isocyanate monomers and oligomers.

Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service