Key Documents
502391
Micro-Splitter Valve (Low Pressure)
10-32 fingertight fittings
About This Item
UNSPSC Code:
41115854
eCl@ss:
32060101
material
PEEK/PTFE
feature
number of ports 3
manufacturer/tradename
Upchurch Scientific, Inc Micro-Splitter P-451
parameter
800 psi max. pressure
fitting
10-32 fingertight fittings
General description
  • Inert polyether ether ketone (PEEK) construction
  • Flow rates as low as 3.5 μL/min (water)
  • 1/4-28 or 10-32 1/16 inch fittings
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)