50192-U
Partisil™ 10 ODS HPLC Column
10 μm particle size, L × I.D. 25 cm × 4.6 mm
Synonym(s):
Partisil™ HPLC Column
About This Item
Recommended Products
Agency
suitable for USP L1
product line
Partisil™
feature
endcapped: no
manufacturer/tradename
Partisil™
extent of labeling
5% Carbon loading
parameter
≤70 °C temp. range
technique(s)
HPLC: suitable
L × I.D.
25 cm × 4.6 mm
surface area
350 m2/g
matrix
octadecyl
matrix active group
C18 (octadecyl) phase
particle size
10 μm
pore size
85 Å
separation technique
reversed phase
Looking for similar products? Visit Product Comparison Guide
Application
- Determination of a new antimalarial drug, benflumetol, in blood plasma by high-performance liquid chromatography.: This study describes the analytical application of the Partisil™ 10 ODS HPLC column for determining benflumetol concentrations in plasma, crucial for pharmacokinetic evaluations (Mansor SM et al., 1996) (Mansor SM et al., 1996).
- High-performance liquid chromatographic separation of platinum complexes containing the cis-1,2-diaminocyclohexane carrier ligand.: Demonstrates the use of Partisil™ 10 ODS HPLC columns in the separation of cisplatin analogs, critical for developing new chemotherapeutic agents (Mauldin SK et al., 1986) (Mauldin SK et al., 1986).
- Separation of retinyl esters by non-aqueous reversed-phase high-performance liquid chromatography.: This research highlights the adaptability of Partisil™ 10 ODS columns to non-aqueous solvents, expanding its applicability in vitamin analysis and nutritional studies (Furr HC et al., 1986) (Furr HC et al., 1986).
- Assay of mixtures of padimate-O and oxybenzone in sunscreen formulations by high-performance liquid chromatography.: Focuses on the role of Partisil™ 10 ODS HPLC columns in ensuring safety and efficacy of sunscreen products by precise quantification of UV filters, contributing significantly to public health safety (Tan HS et al., 1984) (Tan HS et al., 1984).
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service