Skip to Content
Merck
All Photos(1)

Key Documents

49117

Supelco

Silvex®

PESTANAL®, analytical standard

Synonym(s):

2-(2,4,5-Trichlorophenoxy)propionic acid, 2,4,5-TP, Fenoprop, Silvex®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7Cl3O3
CAS Number:
Molecular Weight:
269.51
Beilstein:
1985768
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

CofA

current certificate can be downloaded

packaging

ampule of 25 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

room temp

SMILES string

CC(Oc1cc(Cl)c(Cl)cc1Cl)C(O)=O

InChI

1S/C9H7Cl3O3/c1-4(9(13)14)15-8-3-6(11)5(10)2-7(8)12/h2-4H,1H3,(H,13,14)

InChI key

ZLSWBLPERHFHIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silvex® is a herbicide and a plant growth regulator.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Silvex® may be used as a reference standard in the determination of silvex in water samples and soil samples using high performance liquid chromatography (HPLC) and gas chromatography technique (GC), respectively.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
Silvex is a registered trademark of Silberline Manufacturing Co., Inc.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of 2, 4-dichlorophenoxyacetic acid and silvex in water with high-performance liquid chromatography.
Vaughan W C.
Analytica Chimica Acta, 131, 307-310 (1981)
Residues in apples sprayed with fenoprop.
Cochrane P W, et al.
Canadian Journal of Plant Science , 56(1), 207-210 (1976)
Determinations of Clopyralid, Picloram, and Silvex at Low Concentration in Soils by Calcium Hydroxide? Water Extraction and Gas Chromatography Measurement.
Tan K L, et al.
Journal of Agricultural and Food Chemistry, 44(4), 1135-1143 (1996)
E Herrero-Hernández et al.
Analytica chimica acta, 650(2), 195-201 (2009-09-02)
An extraction-preconcentration procedure based on the use of a molecularly imprinted polymer (MIP) as selective sorbent has been developed for the determination of several phenolic compounds (bisphenol-A, bisphenol-F and 4-nitrophenol) and phenoxyacid herbicides (2,4-D, 2,4,5-T and 2,4,5-TP) in honey samples.
Silvex (2,4,5-TP).
Reviews of environmental contamination and toxicology, 104, 195-202 (1988-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service