Skip to Content
Merck
All Photos(1)

Key Documents

49007

Supelco

BHC (mixture of hexachlorocyclohexanes)

analytical standard, ampule of 500 mg

Synonym(s):

HCH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6Cl6
CAS Number:
Molecular Weight:
290.83
EC Number:
MDL number:
UNSPSC Code:
12000000

grade

analytical standard

packaging

ampule of 500 mg

application(s)

agriculture
environmental

SMILES string

ClC1C(Cl)C(Cl)C(Cl)C(Cl)C1Cl

InChI

1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H

InChI key

JLYXXMFPNIAWKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Pućko et al.
Environmental science & technology, 46(21), 11862-11869 (2012-10-09)
During the summer of 2009, we sampled 14 partially refrozen melt ponds and the top 1 m of old ice in the pond vicinity for α-hexachlorocyclohexane (α-HCH) concentrations and enantiomer fractions (EFs) in the Beaufort Sea. α-HCH concentrations were 3
Fiona Wong et al.
Environmental science and pollution research international, 19(6), 1908-1914 (2012-07-07)
The air-sea gas exchange of alpha-hexachlorocyclohexane (α-HCH) in the Canadian Arctic was estimated using a micrometeorological approach and the commonly used Whitman two-film model. Concurrent shipboard measurements of α-HCH in air at two heights (1 and 15 m) and in
Pernilla Carlsson et al.
Journal of environmental monitoring : JEM, 14(5), 1437-1443 (2012-04-07)
A pilot study is presented evaluating selected chlorinated pesticides as chemical tracers for water masses in a sub-Arctic fjord system (Godthåbsfjord, western Greenland). Polyoxymethylene (POM) based passive water samplers were deployed during summer-autumn 2010. The levels of the analysed chlorinated
Chongguo Tian et al.
Environmental science & technology, 46(4), 2220-2227 (2012-01-21)
Atmospheric outflow of α-HCH from China from 1952 to 2009 was investigated using Chinese Gridded Pesticide Emission and Residue Model (ChnGPERM). The model results show that the outflows via the northeast boundary (NEB, longitude 115-135 °E along 55 °N and
Esha Sharma et al.
European journal of obstetrics, gynecology, and reproductive biology, 161(2), 163-169 (2012-02-09)
Organochlorine pesticides (OCPs) and oxidative stress are reported to be associated with adverse reproductive outcomes. Glutathione S-transferase (GST) is a polymorphic supergene family involved in the detoxification of numerous toxins including OCPs. OCPs are endocrine disrupter and prenatal exposure to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service