Skip to Content
Merck
All Photos(1)

Key Documents

48673

Supelco

Aldrin solution

20 μg/mL in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8Cl6
CAS Number:
Molecular Weight:
364.91
Beilstein:
2336652
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1 mL

concentration

20 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

SMILES string

ClC1=C(Cl)[C@]2(Cl)[C@@H]3[C@@H]4C[C@@H](C=C4)[C@@H]3[C@@]1(Cl)C2(Cl)Cl

InChI

1S/C12H8Cl6/c13-8-9(14)11(16)7-5-2-1-4(3-5)6(7)10(8,15)12(11,17)18/h1-2,4-7H,3H2/t4-,5+,6+,7-,10+,11-

InChI key

QBYJBZPUGVGKQQ-SJJAEHHWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glenn A Rowland et al.
Environmental pollution (Barking, Essex : 1987), 159(5), 1076-1084 (2011-03-15)
Organochlorine (OC) contaminants are transported to the Polar Regions, where they have the potential to bioaccumulate, presenting a threat to the health of wildlife and indigenous communities. They deposit onto snowpack during winter, and accumulate until spring, when they experience
Lisa Tröbs et al.
Environmental science and pollution research international, 18(4), 547-555 (2010-10-05)
3-Chlorocarbazole and 3,6-dichlorocarbazole were isolated from Bavarian soils. The stereospecific formation of the isomers of these chlorinated carbazols can be explained by quantum mechanical calculations using the DFT method. It was shown that chlorination of carbazole and 3-chlorocarbazole respectively is
Le Kang et al.
Epidemiology (Cambridge, Mass.), 21 Suppl 4, S58-S63 (2011-04-01)
Understanding the health effects associated with environmental chemicals is challenging when individuals have concentrations at or below the laboratory limits of detection as well as when the values may round to zero or are presented in the form of 0
Xiao-Yuan Tao et al.
Molecular ecology, 21(17), 4371-4385 (2012-04-21)
Cotton plants accumulate phytotoxins, including gossypol and related sesquiterpene aldehydes, to resist insect herbivores and pathogens. To counteract these defensive plant secondary metabolites, cotton bollworms (Helicoverpa armigera) elevate their production of detoxification enzymes, including cytochrome P450 monooxygenases (P450s). Besides their
Xiaomin Li et al.
Journal of chromatography. A, 1277, 69-75 (2013-01-22)
Identification and quantification of related-structure impurity is a research focus in the purity assessment of organic compounds. Determination of the purity value and uncertainty assessment are also important in the metrological research. A method for the determination of related-structure impurity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service