Skip to Content
Merck
All Photos(1)

Key Documents

48639

Supelco

1,2-Dichlorobenzene solution

certified reference material, 200 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Cl2
CAS Number:
Molecular Weight:
147.00
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

certified reference material

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-30°C

SMILES string

Clc1ccccc1Cl

InChI

1S/C6H4Cl2/c7-5-3-1-2-4-6(5)8/h1-4H

InChI key

RFFLAFLAYFXFSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

50.0 °F

Flash Point(C)

10 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eranda Maligaspe et al.
The journal of physical chemistry. A, 114(1), 268-277 (2009-11-26)
Singlet-singlet energy transfer in self-assembled via axial coordination of imidazole-appended (at different positions of one of the meso-phenyl entities) free-base tetraphenylporphyrin, H(2)PIm, to either zinc phthalocyanine, ZnPc, or zinc naphthalocyanine, ZnNc, dyads is investigated in noncoordinating solvents, o-dichlorobenzene and toluene
Bimalendu Adhikari et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(41), 11488-11496 (2011-09-29)
The N-terminally pyrene-conjugated oligopeptide, Py-Phe-Phe-Ala-OMe, (Py=pyrene 1-butyryl acyl) forms transparent, stable, supramolecular fluorescent organogels in various organic solvents. One of these organogels was thoroughly studied using various techniques including transmission electron microscopy (TEM), field-emission scanning electron microscopy (FESEM), atomic force
Ivan Guryanov et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(46), 12837-12845 (2009-10-23)
The effect of microwave (MW) irradiation and ionic liquids (IL) on the cycloaddition of azomethine ylides to [60]fullerene has been investigated by screening the reaction protocol with regard to the IL medium composition, the applied MW power, and the simultaneous
Eric Vejerano et al.
Environmental science & technology, 45(2), 589-594 (2010-12-09)
Previous studies have shown environmentally persistent free radicals (EPFRs) form when chlorine- and hydroxy-substituted benzenes chemisorb on Cu(II)O-containing surfaces under postcombustion conditions. This paper reports the formation of EPFRs on silica particles containing 5% Fe(III)(2)O(3). The EPFRs are formed by
Sungmin Chin et al.
Chemosphere, 75(9), 1206-1209 (2009-03-10)
This study examined the catalytic oxidation of 1,2-dichlorobenzene on V(2)O(5)/TiO(2) nanoparticles. The V(2)O(5)/TiO(2) nanoparticles were synthesized by the thermal decomposition of vanadium oxytripropoxide and titanium tetraisopropoxide. The effects of the synthesis conditions, such as the synthesis temperature and precursor heating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service