Skip to Content
Merck
All Photos(1)

Key Documents

48609

Supelco

Tetrachloroethene solution

certified reference material, 200 μg/mL in methanol

Synonym(s):

Tetrachloroethene solution, Perchloroethylene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental
petroleum

format

single component solution

storage temp.

2-30°C

InChI

1S/C2Cl4/c3-1(4)2(5)6

InChI key

CYTYCFOTNPOANT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrachloroethene is a colorless liquid having an ethereal odor, which can find applications as a chemical intermediate and dry cleaning agent.[1] It can also undergo reductive dehalogenation in the presence of an anaerobic bacterium to yield cis-1,2-dichloroethene.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hunter's Diseases of Occupations, Tenth Edition (2010)
Isolation and characterization of a facultatively aerobic bacterium that reductively dehalogenates tetrachloroethene to cis-1, 2-dichloroethene
Sharma.KP and Mccarty.LP
Applied and Environmental Microbiology, 62, 761-765 (1996)
Koen G Zuurbier et al.
Journal of contaminant hydrology, 147, 1-13 (2013-02-26)
Groundwater systems are increasingly used for seasonal aquifer thermal energy storage (SATES) for periodic heating and cooling of buildings. Its use is hampered in contaminated aquifers because of the potential environmental risks associated with the spreading of contaminated groundwater, but
Jan E Storm et al.
Environmental research, 122, 88-97 (2013-03-05)
In many cities, dry cleaners using perchloroethylene are frequently located in multifamily residential buildings and often cause elevated indoor air levels of perchloroethylene throughout the building. To assess individual perchloroethylene exposures associated with co-located dry cleaners, we measured perchloroethylene in
Stephen G Whittaker et al.
Journal of environmental health, 75(10), 14-22 (2013-07-19)
Workers in the dry cleaning industry are exposed to a variety of harmful solvents, and poor work practices can result in extensive environmental contamination. Of particular concern is perchloroethylene (PERC), which is the most commonly used cleaning solvent. This chlorinated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service