Skip to Content
Merck
All Photos(1)

Key Documents

48599

Supelco

2,3,7,8-Tetrachlorodibenzo-p-dioxin solution

analytical standard

Synonym(s):

2,3,7,8-TCDD

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12000000

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

storage temp.

2-8°C

InChI

1S/C12H4Cl4O2/c13-5-1-9-10(2-6(5)14)18-12-4-8(16)7(15)3-11(12)17-9/h1-4H

InChI key

HGUFODBRKLSHSI-UHFFFAOYSA-N

General description

2,3,7,8-Tetrachlorodibenzo-p-dioxin is a highly distributed environmental contaminant[1], released by industrial activities, which can act like as a reproductive toxicant and endocrine disruptor in all vertebrates.[2]
2,3,7,8-Tetrachlorodibenzo-p-dioxin is the most potent congener of dioxins and dioxin-like chemicals, which can exert adverse effects in laboratory animals used in testing.[3]

Application

2,3,7,8-Tetrachlorodibenzo-p-dioxin solution has been used as standard in investigating the effect of 2,3,7,8-Tetrachlorodibenzo-p-dioxin to offsprings of fish, using high performance gas chromatography coupled with high performance mass spectrometry (HPGC-HPMS).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

E C Rada et al.
Chemosphere, 110, 53-61 (2014-06-02)
Atmospheric depositions of polychlorinated dibenzo-p-dioxins and dibenzofurans (PCDD/Fs) and polychlorinated biphenyls (PCBs) were monitored at three sites in the vicinity of a steel making plant, located in an Italian alpine valley. A high variability in the deposition of PCDD/Fs was
K A Solak et al.
Toxicology, 311(3), 124-134 (2013-07-23)
The aryl hydrocarbon receptor (AhR) is involved in a wide variety of biological and toxicological responses, including neuroendocrine signaling. Due to the complexity of neuroendocrine pathways in e.g. the hypothalamus and pituitary, there are limited in vitro models available despite
Filomena Fiorito et al.
Toxicology, 290(2-3), 258-270 (2011-10-22)
The administration of 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD) to a variety of cultured cells may alter their ability to proliferate and die. In a previous study we demonstrated that TCDD induced proliferation in Madin-Darby Bovine Kidney (MDBK) cells where no signs of apoptosis
Maria Herlin et al.
Toxicology and applied pharmacology, 273(1), 219-226 (2013-09-17)
Bone is a target for high affinity aryl hydrocarbon receptor (AHR) ligands, such as dioxins. Although bone morphology, mineral density and strength are sensitive endpoints of dioxin toxicity, less is known about effects on bone microarchitecture and material properties. This
Tomoki Takeda et al.
Molecular pharmacology, 85(1), 74-82 (2013-10-18)
Our previous studies have shown that treatment of pregnant rats with 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD; 1 μg/kg) at gestational day (GD) 15 reduces the pituitary synthesis of luteinizing hormone (LH) during the late fetal and early postnatal period, leading to the imprinting

Questions

1–4 of 4 Questions  
  1. Can I get pure 2,3,7,8-Tetrachlorodibenzo-p-dioxin from Sigma-Aldrich?

    1 answer
    1. No; Sigma-Aldrich has no source for pure 2,3,7,8-Tetrachlorodibenzo-p-dioxin.  It is only available as Supelco 48599: 2,3,7,8-TCDD, 10 μg/mL in toluene, ampule of 1mL.

      Helpful?

  2. What is the molecular weight of 2,3,7,8-Tetrachlorodibenzo-p-dioxin, Product 48599?

    1 answer
    1. The molecular weight of 2,3,7,8-TCDD is 321.97 Da.

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. Is Product No. 48599, 2,3,7,8-Tetrachlorodibenzo-p-dioxin solution, appropriate for use in tissue culture studies?

    1 answer
    1. The 2,3,7,8-Tetrachlorodibenzo-p-dioxin solution (10 μg/mL in toluene), is sold as 1mL in a flame sealed ampule.  It is made to be used as a chemical standard with US EPA drinking and wastewater analytical methods.  We have no knowledge of its use in tissue culture.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service