Skip to Content
Merck
All Photos(1)

Key Documents

48574

Supelco

Dibenz[a,h]anthracene

analytical standard

Synonym(s):

1,2:5,6-Dibenzanthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14
CAS Number:
Molecular Weight:
278.35
Beilstein:
1912416
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

vial of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

524 °C (lit.)

mp

262-265 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

c1ccc2c(c1)ccc3cc4c(ccc5ccccc45)cc23

InChI

1S/C22H14/c1-3-7-19-15(5-1)9-11-17-14-22-18(13-21(17)19)12-10-16-6-2-4-8-20(16)22/h1-14H

InChI key

LHRCREOYAASXPZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Shou et al.
Archives of biochemistry and biophysics, 328(1), 201-207 (1996-04-01)
The metabolic activation of the potent carcinogen dibenzo[a,h]anthracene (DB[a,h]A) was investigated with recombinant human cytochrome P450 enzymes 1A2, 2B6, 2C8, 2C9, 2E1, 3A3, 3A4, and 3A5 expressed in hepatoma G2 cells and with 14 different human liver microsomes. Three dihydrodiols
A Perico et al.
Archives of environmental health, 56(6), 506-512 (2002-04-18)
In this study, the authors evaluated exposure to airborne polycyclic aromatic hydrocarbons (PAHs) in workers exposed to exhaust gas from cars, and they assessed the efficiency of urinary 1-hydroxypyrene as an indicator of exposure to pyrene and PAHs. The authors
A L Juhasz et al.
Letters in applied microbiology, 30(5), 396-401 (2000-05-03)
The ability of Stenotrophomonas maltophilia strain VUN 10,003 to degrade and detoxify high molecular weight polycyclic aromatic hydrocarbons (PAHs) was evaluated in a basal liquid medium. Using high cell density inocula of strain VUN 10,003, the concentration of pyrene, fluoranthene
B Z Zhong et al.
Mutation research, 326(2), 147-153 (1995-02-01)
Benz[a]anthracene (BA), dibenz[a,h]anthracene (DBA) and dibenzo[a,i]pyrene (DBP) are polycyclic aromatic hydrocarbons (PAHs) found in incomplete combustion products of fossil fuels, coal tar, and other organic materials. Workers in related industries may be exposed to these chemicals by inhalation. The information
P Sartorelli et al.
Toxicology and industrial health, 17(1), 17-21 (2002-05-15)
To understand the dermal uptake of chemicals bound to soil and dust, information on the neat substance is helpful but does not seem sufficient. Because of its content of polycyclic aromatic hydrocarbons (PAHs) coal is suspected to be carcinogenic. However

Articles

This application note describes the fast and efficient separation of the EU’s list of 15 + 1 polynuclear aromatic hydrocarbons (PAHs) using Ascentis® Express PAH HPLC column.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service