Skip to Content
Merck
All Photos(1)

Key Documents

48094

Supelco

Phenanthrene-d10 solution

certified reference material, 2000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

c21c3c(c(c(c2c(c(c(c1[2H])[2H])[2H])[2H])[2H])[2H])c(c(c(c3[2H])[2H])[2H])[2H]

InChI

1S/C14H10/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)13/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D

InChI key

YNPNZTXNASCQKK-LHNTUAQVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

Identification and quantification of the most common extractables originating from single-use equipment by GC/MS and an Extractables and Leachables Screening Standard.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service