Skip to Content
Merck
All Photos(1)

Key Documents

48083

Supelco

4-Bromofluorobenzene solution

certified reference material, 2000 μg/mL in methanol

Synonym(s):

1-Bromo-4-fluorobenzene solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4BrF
CAS Number:
Molecular Weight:
175.00
EC Number:
MDL number:
UNSPSC Code:
12000000
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 624,8010,8020,501.3,524.1,524.2

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type surrogate

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)Br

InChI

1S/C6H4BrF/c7-5-1-3-6(8)4-2-5/h1-4H

InChI key

AITNMTXHTIIIBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
4-Bromofluorobenzene solution has been used as an internal standard in the determination of the carcinogen, 1,4-dioxane in household detergents and cleaners using gas chromatography coupled with mass spectrometry method (GC-MS).[1] It has also been used as an internal standard in the estimation of pollutants in environmental samples using pervaporation technique and gas chromatography coupled with mass spectrometry (GC-MS).[2]

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup - Solvent

Flash Point(C)

11 °C - closed cup - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of non-covalent interactions between organic compounds and humic substances on analytical pervaporation
Mishima.S and Nakagawa.T
Journal of Membrane Science , 228(1), 1-4 (2004)
Determination of 1, 4-dioxane in household detergents and cleaners
Tanabe A, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 91(2), 439-444 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service