Skip to Content
Merck
All Photos(1)

Key Documents

442804

Supelco

Tokuthion

technical grade, pkg of 50 mg

Synonym(s):

Prothiofos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15Cl2O2PS2
CAS Number:
Molecular Weight:
345.25
Beilstein:
1998314
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

technical grade

packaging

pkg of 50 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCCSP(=S)(OCC)Oc1ccc(Cl)cc1Cl

InChI

1S/C11H15Cl2O2PS2/c1-3-7-18-16(17,14-4-2)15-11-6-5-9(12)8-10(11)13/h5-6,8H,3-4,7H2,1-2H3

InChI key

FITIWKDOCAUBQD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tokuthion may be used as a reference standard for the analysis of tokuthion, an organic contaminant in continental bottled water by stir bar sorptive extraction followed by gas chromatography coupled with tandem mass spectrometry (SBSE-GC-MS/MS).

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Continental bottled water assessment by stir bar sorptive extraction followed by gas chromatography-tandem mass spectrometry (SBSE-GC-MS/MS).
Guart A, et al.
Environmental Science and Pollution Research International, 21(4), 2846-2855 (2014)
Takahiro Miyo et al.
Journal of economic entomology, 95(5), 871-877 (2002-10-31)
To investigate the genetic basis of cross-resistance to insecticides, we conducted genetic analyses of resistance to three organophosphate insecticides, malathion, prothiophos, and fenitrothion. After isofemale lines resistant and susceptible to all of the three organophosphates had been screened from natural
[Hygienic regulation of the insecticide prothiophos in reservoir waters].
N M Demidenko et al.
Gigiena i sanitariia, (5)(5), 92-93 (1988-05-01)
[Effect of the pesticide tocuthion on the activity of microsomal cytochrome P-450-dependent monooxygenases in the rat liver].
N M Demidenko et al.
Gigiena truda i professional'nye zabolevaniia, (1)(1), 45-46 (1990-01-01)
M Sakata et al.
Journal of toxicology. Clinical toxicology, 37(3), 327-332 (1999-06-29)
A 63-year-old woman was admitted to a local hospital after the ingestion of 40% prothiofos preparation (Tokuthion) 370 mL. Gastric lavage was performed and cathartics, active charcoal, diuretics, atropine sulfate, and pralidoxime were administered. Serum cholinesterase activity was 1.3 IU/L

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service