Skip to Content
Merck
All Photos(1)

Key Documents

442605

Supelco

Hexabromobenzene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6Br6
CAS Number:
Molecular Weight:
551.49
Beilstein:
1912586
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

>300 °C (lit.)

application(s)

environmental

format

neat

storage temp.

room temp

SMILES string

Brc1c(Br)c(Br)c(Br)c(Br)c1Br

InChI

1S/C6Br6/c7-1-2(8)4(10)6(12)5(11)3(1)9

InChI key

CAYGQBVSOZLICD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Koss et al.
Toxicology letters, 14(1-2), 69-77 (1982-11-01)
To study the metabolic fate of hexabromobenzene (HBB) in female rats, the substance was administered in oral doses of 16.6 mg/kg body weight every other day for 2 weeks and the animals' excreta were examined for metabolites. Unchanged HBB, pentabromobenzene
K D Courtney et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 19(1), 83-94 (1984-02-01)
HBB (hexabromobenzene) and HFB (hexafluorobenzene) were tested for their teratogenic potential in CD-1 mice. HBB and HFB were administered to pregnant mice from the 6th to the 16th day of gestation by gastric intubation. Neither HBB nor HFB were teratogenic
Lewis T Gauthier et al.
Environmental science & technology, 43(2), 312-317 (2009-02-26)
The production and use of nonpolybrominated diphenyl ether (non-PBDE), brominated flame retardant (BFR) alternatives have been on the rise, although their assessment in environmental samples is largely understudied. In the present study, several non-PBDE BFRs were found in the egg
Amina Salamova et al.
Environmental science & technology, 45(20), 8698-8706 (2011-09-29)
Air (vapor and particle) and precipitation samples were collected at five sites (two urban, one rural, and two remote) around the Great Lakes during 2005-2009 as a part of the Integrated Atmospheric Deposition Network (IADN). The concentrations of polybrominated diphenyl
Jiang-Ping Wu et al.
Environment international, 37(1), 210-215 (2010-10-19)
With the phaseout of brominated flame retardants (BFRs) polybrominated diphenyl ethers (PBDEs), some non-PBDE BFRs have prompted to be alternatives to the discontinued PBDEs. To assess the bioaccumulation potential of these chemicals, field bioaccumulation factors (BAFs) for several non-PBDE BFRs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service