Skip to Content
Merck
All Photos(1)

Key Documents

442481

Supelco

Benzyl alcohol

analytical standard

Synonym(s):

Benzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
878307
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
39023110
PubChem Substance ID:

grade

analytical standard

vapor density

3.7 (vs air)

vapor pressure

13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

autoignition temp.

817 °F

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.539 (lit.)

bp

203-205 °C (lit.)

mp

−16-−13 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

OCc1ccccc1

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzyl Alcohol is a type of aromatic alcohol mainly used in cosmetic products and preservatives. It is also used as a solvent and viscosity-decreasing agent.

Application

Benzyl alcohol has been used as a reference standard for the analysis of the analyte in scented consumer goods by gas chromatography-mass spectrometry (GC-MS) as well as gas chromatography-olfactometry (GC-O). It has been also used as a reference standard for the determination of the analyte in “suspected” skin allergen mixture and in fragrance allergens by GC-MS. It may be used as a reference standard for the identification of the analyte in bio-oils obtained from pyrolysis of various feedstock and in Camellia sinensis (L.) Kuntze black tea by GC-MS.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

213.8 °F - DIN 51758

Flash Point(C)

101 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of odour compounds from scented consumer products using gas chromatography-mass spectrometry and gas chromatography-olfactometry
Bartsch J, et al.
Analytica Chimica Acta, 904, 98-106 (2016)
A simple procedure for chromatographic analysis of bio-oils from pyrolysis
Undri A, et al.
Journal of Analytical and Applied Pyrolysis, 114, 208-221 (2015)
Pressurized liquid extraction-gas chromatography-mass spectrometry analysis of fragrance allergens, musks, phthalates and preservatives in baby wipes
Celeiro M, et al.
Journal of Chromatography A, 1384, 9-21 (2015)
Analysis of essential oils and fragrances with a new generation of highly inert gas chromatographic columns coated with ionic liquids
Cagliero C, et al.
Journal of Chromatography A, 1495, 64-75 (2017)
Final report on the safety assessment of Benzyl Alcohol, Benzoic Acid, and Sodium Benzoate
NB
International Journal of Toxicology, 20, 23-50 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service