Skip to Content
Merck
All Photos(1)

Documents

442250

Supelco

1,4-Dinitrobenzene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(NO2)2
CAS Number:
Molecular Weight:
168.11
Beilstein:
1105828
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

183.4 °C/34 mmHg (lit.)

mp

170-173 °C (lit.)

density

1.625 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

room temp

SMILES string

[O-][N+](=O)c1ccc(cc1)[N+]([O-])=O

InChI

1S/C6H4N2O4/c9-7(10)5-1-2-6(4-3-5)8(11)12/h1-4H

InChI key

FYFDQJRXFWGIBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methemoglobinemia due to occupational exposure to dinitrobenzene--Ohio, 1986.
MMWR. Morbidity and mortality weekly report, 37(22), 353-355 (1988-06-10)
N Kessler et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 10(12), 1435-1442 (1996-10-01)
Monoclonal antibodies have been elicited and selected after injection of crystals of 1,4-dinitrobenzene (1,4-DNB) and cholesterol monohydrate in mice. The reactivity of some of these antibodies to 1,4-DNB crystals, cholesterol monohydrate crystals, and other solid substrates has been characterized. Two
Akio Kamimura et al.
Molecules (Basel, Switzerland), 17(5), 4782-4790 (2012-04-27)
S(RN)1-type coupling adducts are readily prepared by the reaction between a-sulfonylesters or a-cyanosulfones and gem-dinitro compounds in ionic liquids. The reactions progress smoothly and recovered ionic liquids can be used for several iterations, as long as they are washed with
Yanyan Zhao et al.
Se pu = Chinese journal of chromatography, 29(9), 885-889 (2012-01-12)
A set of 14 model compounds were selected to evaluate the retention properties of two silica-based cyclodextrin (CD) bonded stationary phases with different spacers (Click Alkyl-CD and Click OEG-CD) under reversed-phase liquid chromatographic (RPLC) mode. The gradient calculation method and
Silvia M Barolo et al.
The Journal of organic chemistry, 71(22), 8493-8499 (2006-10-27)
The photostimulated intramolecular ortho-arylation reactions of bromoarenes linked with pendant phenoxy containing N-substituted tetrahydroisoquinolines in liquid ammonia afforded aporphine (54-82% yield) alkaloid derivatives via SRN1 reactions. This strategy was extended for the first time to the synthesis of a homoaporphine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service