Skip to Content
Merck
All Photos(1)

Key Documents

43266

Supelco

Diniconazole

PESTANAL®, analytical standard, E-Isomer

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€166.00

€166.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
250 MG
€166.00

About This Item

Empirical Formula (Hill Notation):
C15H17Cl2N3O
CAS Number:
Molecular Weight:
326.22
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€166.00


Estimated to ship on30 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

quality

E-Isomer

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water

mp

152-158 °C

format

neat

SMILES string

OC(C(C)(C)C)/C(N1N=CC=C1)=C\C2=CC=C(Cl)C=C2Cl

InChI

1S/C15H17Cl2N3O/c1-15(2,3)14(21)13(20-9-18-8-19-20)6-10-4-5-11(16)7-12(10)17/h4-9,14,21H,1-3H3/b13-6+

InChI key

FBOUIAKEJMZPQG-AWNIVKPZSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service