Skip to Content
Merck
All Photos(1)

Documents

42263

Supelco

Ethyl 4-formamidobenzoate

reference material

Synonym(s):

p-Formamidobenzoic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO3
CAS Number:
Molecular Weight:
193.20
MDL number:
UNSPSC Code:
41116107

grade

reference material

API family

benzocaine

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

SMILES string

O=C(OCC)C1=CC=C(NC([H])=O)C=C1

InChI

1S/C10H11NO3/c1-2-14-10(13)8-3-5-9(6-4-8)11-7-12/h3-7H,2H2,1H3,(H,11,12)

InChI key

XXNAVWORVSFFAD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service