Skip to Content
Merck
All Photos(1)

Key Documents

42230

Supelco

Meranzin hydrate

analytical standard

Synonym(s):

8-[(2S)-2,3-Dihydroxy-3-methylbutyl]-7-methoxy-2H-1-benzopyran-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€516.00

€516.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€516.00

About This Item

Empirical Formula (Hill Notation):
C15H18O5
CAS Number:
Molecular Weight:
278.30
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€516.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

optical activity

[α]/D -32.0 to -26.0°, c = 0.11 in chloroform

shelf life

limited shelf life, expiry date on the label

storage temp.

−20°C

SMILES string

O=C1C=CC2=CC=C(OC)C(C[C@H](C(O)(C)C)O)=C2O1

InChI

1S/C15H18O5/c1-15(2,18)12(16)8-10-11(19-3)6-4-9-5-7-13(17)20-14(9)10/h4-7,12,16,18H,8H2,1-3H3/t12-/m0/s1

InChI key

KGGUASRIGLRPAX-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adriana Arigò et al.
Journal of chromatography. A, 1649, 462183-462183 (2021-05-27)
In the European Union, cosmetic products are regulated by the Cosmetics Regulation (EC) No. 1223/2009. The Analytical Working Group of the International Fragrance Association (IFRA) suggested that a new sensitive analytical method is needed to determine psoralens in finished cosmetic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service