Skip to Content
Merck
All Photos(1)

Key Documents

41651

Supelco

L-Rhamnose monohydrate

analytical standard

Synonym(s):

6-Deoxy-L-mannose, L(+)Rhamnopyranose

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€62.10

€62.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€62.10

About This Item

Empirical Formula (Hill Notation):
C6H12O5 · H2O
CAS Number:
Molecular Weight:
182.17
Beilstein:
3912643
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.24

€62.10


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

optical activity

[α]/D 8.0±0.5°, 2 hr, c = 5 in H2O

shelf life

limited shelf life, expiry date on the label

impurities

9.0-11.0% waer

application(s)

food and beverages

format

neat

SMILES string

O.C[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C6H12O5.H2O/c1-2-3(7)4(8)5(9)6(10)11-2;/h2-10H,1H3;1H2/t2-,3-,4+,5+,6+;/m0./s1

InChI key

BNRKZHXOBMEUGK-NRBMBCGPSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service