Skip to Content
Merck
All Photos(1)

Key Documents

41467

Supelco

Harmane

analytical standard

Synonym(s):

1-Methyl-9H-pyrido[3,4-b]indole, 2-Methyl-β-carboline, Aribine

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€94.50

€94.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€94.50

About This Item

Empirical Formula (Hill Notation):
C12H10N2
CAS Number:
Molecular Weight:
182.22
Beilstein:
143898
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€94.50


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

235-238 °C (lit.)
235-238 °C

SMILES string

Cc1nccc2c3ccccc3[nH]c12

InChI

1S/C12H10N2/c1-8-12-10(6-7-13-8)9-4-2-3-5-11(9)14-12/h2-7,14H,1H3

InChI key

PSFDQSOCUJVVGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

I1 imidazoline binding site agonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service