Skip to Content
Merck
All Photos(1)

Key Documents

41103

Supelco

1,3,5-Trimethylbenzene solution

certified reference material, 5000 μg/mL in methanol

Synonym(s):

Mesitylene solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C9H12/c1-7-4-8(2)6-9(3)5-7/h4-6H,1-3H3

InChI key

AUHZEENZYGFFBQ-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Cushion et al.
Dalton transactions (Cambridge, England : 2003), (9)(9), 1632-1635 (2009-05-08)
The new sterically encumbered facially coordinating N(3)-donor ligand cis,cis-1,3,5-tris(mesitylideneimino)cyclohexane (L1) has been synthesised. Reaction with [Cu(NCCH(3))(4)]PF(6) gives [Cu(L1)NCCH(3)]PF(6) (1), the bound acetonitrile being labile and readily replaced by CO to yield [Cu(L1)CO]PF(6) (2); both 1 and 2 have been structurally
M Prager et al.
Physical chemistry chemical physics : PCCP, 7(13), 2587-2593 (2005-09-29)
Mesitylene can be stabilized at He temperature in three solid phases of so far unknown crystal structures. Rotational tunneling of methyl groups is based on rotational potentials and used to characterize structural aspects. In phase III found after the first
Dorota Wiaderna et al.
International journal of occupational medicine and environmental health, 15(4), 385-392 (2003-03-01)
Trimetylbenzene isomers: pseudocumene, hemimellitene and mesitylene, are major components of numerous commercial solvents and high-grade fuels. In our earlier research on rats we have proved that inhalation exposure to pseudocumene or hemimellitene at concentrations close to the MAC value results
Natalie D Coombs et al.
Dalton transactions (Cambridge, England : 2003), (3)(3), 332-337 (2008-04-17)
Sterically encumbered amido ligands based on a 1,8-diarylcarbazol-9-yl backbone have been investigated as electronically distinct alternatives to the widely-used terphenyl ligand class in the stabilization of low-coordinate metal complexes, and structurally characterized for the first time. While 1,8-diphenylcarbazol-9-yl derivatives are
Sanyog Sharma et al.
Organic & biomolecular chemistry, 11(4), 654-661 (2012-12-12)
Two mesitylene based probes, having catechol/phenol units in conjunction to the Schiff base have been synthesized. The probe with a catechol unit can be used to sense and discriminate between F(-) and CN(-) through different colorimetric and fluorimetric responses using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service