Skip to Content
Merck
All Photos(1)

Key Documents

41097

Supelco

Dibromomethane solution

5000 μg/mL in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

analytical standard

CofA

certificate of analysis is enclosed in each package.

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

BrCBr

InChI

1S/CH2Br2/c2-1-3/h1H2

InChI key

FJBFPHVGVWTDIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glenn A Marsch et al.
Chemical research in toxicology, 17(1), 45-54 (2004-01-20)
The dihalomethane CH(2)Cl(2) is an industrial solvent of potential concern to humans because of its potential genotoxicity and carcinogenicity. To characterize DNA damage by dihalomethanes, a rapid DNA digestion under acidic conditions was developed to identify alkali labile DNA-dihalomethane nucleoside
Marie E Archbold et al.
Rapid communications in mass spectrometry : RCM, 19(3), 337-342 (2005-01-13)
A pre-concentration system has been validated for use with a gas chromatography/mass spectrometry/isotope ratio mass spectrometer (GC/MS/IRMS) to determine ambient air (13)C/(12)C ratios for methyl halides (MeCl and MeBr) and chlorofluorocarbons (CFCs). The isotopic composition of specific compounds can provide
D A Whittington et al.
Biochemistry, 40(12), 3476-3482 (2001-04-12)
To investigate the role of protein cavities in facilitating movement of the substrates, methane and dioxygen, in the soluble methane monooxygenase hydroxylase (MMOH), we determined the X-ray structures of MMOH from Methylococcus capsulatus (Bath) cocrystallized with dibromomethane or iodoethane, or
Andrey Shalit et al.
Physical chemistry chemical physics : PCCP, 14(40), 13989-13996 (2012-09-20)
Single shot time resolved four wave mixing is a powerful tool for the acquisition of dynamic and spectroscopic data from molecules susceptible to bleaching or other photo-induced damage. We add polarization dependence to single shot methods, and demonstrate how magic
Aley G Kalapila et al.
Mutation research, 684(1-2), 35-42 (2009-11-28)
The primary function of O(6)-alkylguanine-DNA alkyltransferase (AGT) is to maintain genomic integrity in the face of damage by both endogenous and exogenous alkylating agents. However, paradoxically, bacterial and mammalian AGTs have been shown to increase cytotoxicity and mutagenicity of dihaloalkanes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service