Skip to Content
Merck
All Photos(1)

Key Documents

40250-U

Supelco

2-Methylphenol solution

certified reference material, 5000 μg/mL in methanol

Synonym(s):

o-Cresol solution, 2-Methylphenol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Agency

EPA TO-8
NIOSH 2001

product line

TraceCERT®

feature

standard type calibration

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C7H8O/c1-6-4-2-3-5-7(6)8/h2-5,8H,1H3

InChI key

QWVGKYWNOKOFNN-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia Fustinoni et al.
Journal of occupational and environmental hygiene, 4(1), 1-9 (2006-12-13)
The characteristics of urinary o-cresol (o-C) and urinary toluene (TOL-U) as biomarkers of occupational exposure to toluene were comparatively evaluated. One hundred healthy male rotogravure printing workers and 161 male and female control subjects were studied. Personal exposure to airborne
Hua-Wei Chen et al.
Water research, 41(10), 2069-2078 (2007-04-10)
Platinum deposited TiO(2) films were prepared on quartz substrates by dip-coating process for the photodecomposition of o-cresol. The characteristics of Pt/TiO(2) and the temporal behavior of o-cresol decomposition by Pt/TiO(2) photocatalysis under visible light irradiation were investigated. Platinum deposited on
Y Abdollahi et al.
Environmental technology, 33(10-12), 1183-1189 (2012-08-04)
The effective removal of o-cresol is currently both an environmental and economic challenge. ZnO is not only an efficient photocatalyst but is also cost effective, as its photoabsorption can extend from the ultraviolet (UV) to the visible range thereby allowing
Osamu Inoue et al.
Industrial health, 46(4), 318-325 (2008-08-22)
This study was initiated to evaluate o-cresol and benzylmercapturic acid in urine in comparison with other biomarkers, as tools to estimate the intensity of occupational exposure to toluene at low levels. In total, 108 solvent exposed workers (engaged in tape
Johanna M Dela Cruz et al.
Journal of mass spectrometry : JMS, 42(2), 178-186 (2006-12-19)
The influence shaped femtosecond laser pulses have on molecular photofragmentation and ionization, coupled with the intrinsic sensitivity of mass spectrometry, results in a powerful tool for fast, accurate, reproducible and quantitative isomeric identification. Complex phase functions are introduced to enhance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service