Skip to Content
Merck
All Photos(1)

Key Documents

40083

Supelco

Tetrachloroethene solution

certified reference material, 5000 μg/mL in methanol

Synonym(s):

Perchloroethylene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental
petroleum

format

single component solution

storage temp.

2-8°C

InChI

1S/C2Cl4/c3-1(4)2(5)6

InChI key

CYTYCFOTNPOANT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Julie C Chambon et al.
Biotechnology and bioengineering, 110(1), 1-23 (2012-08-29)
Reductive dechlorination is a major degradation pathway of chlorinated ethenes in anaerobic subsurface environments, and reactive kinetic models describing the degradation process are needed in fate and transport models of these contaminants. However, reductive dechlorination is a complex biological process
Jan E Storm et al.
Environmental research, 122, 88-97 (2013-03-05)
In many cities, dry cleaners using perchloroethylene are frequently located in multifamily residential buildings and often cause elevated indoor air levels of perchloroethylene throughout the building. To assess individual perchloroethylene exposures associated with co-located dry cleaners, we measured perchloroethylene in
Stephen G Whittaker et al.
Journal of environmental health, 75(10), 14-22 (2013-07-19)
Workers in the dry cleaning industry are exposed to a variety of harmful solvents, and poor work practices can result in extensive environmental contamination. Of particular concern is perchloroethylene (PERC), which is the most commonly used cleaning solvent. This chlorinated
Funda Cengiz-Callioglu et al.
Journal of nanoscience and nanotechnology, 13(7), 4727-4735 (2013-08-02)
This paper discusses the effects of 1,1,2,2 tetrachlorethylen (TCE) non-solvent addition on the independent (electrical conductivity, dielectric constant, surface tension and the theological properties of the solution etc.) and dependent parameters (number of Taylor cones per square meter (NTC/m2), spinning
Chien-Li Lee et al.
Journal of the Air & Waste Management Association (1995), 62(12), 1443-1448 (2013-02-01)
Microwave (MW) is applied to enhance perchloroethylene (PCE) or pentachlorophenol (PCP) removal using zero-valent iron (ZVI; Fe(0)) as the dielectric medium. ZVI has a much higher dielectric loss factor (39.5) than other media; it is capable of absorbing MW radiation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service